BrandTech™ BRANDplates immunoGrade™ 96-Well Microplates Available on GSA/VA Contract for Federal Government customers only.

An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays

Manufacturer: BrandTech™ 781729

 View more versions of this product

Catalog No. 13-882-225

  Check Availability
Add to Cart

    Specifications

    Lid Yes,Yes
    For Use With (Application) For the majority of standard ELISAs as well as solid phase immunoassays
    Format 96-well,96-well
    Binding Property High,High
    Material Polystyrene,Polystyrene
    No. of Wells 96
    Sterile No
    Type 96-Well Microplate,96-Well Microplate
    Volume (Metric) 350μL,350μL
    Well Design Round
    Closure Yes
    Color Black
    Wells 96
    Well Shape Round

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit