BrandTech™ BRANDplates lipoGrade™ 96-Well Microplates Available on GSA/VA Contract for Federal Government customers only.

Immunoassay plate optimized for the immobilization of hydrophobic molecules

Manufacturer: BrandTech™ 781851

 View more versions of this product

Catalog No. 13-882-243

  Check Availability
Add to Cart

    Specifications

    Barcode Yes
    For Use With (Application) Solid phase with hydrophobic molecules and liquid phase with hydrophilic molecules
    Binding Property High
    Material Polystyrene
    Bottom F-bottom
    Sterile No
    Type 96-Well Assay Microplate
    Certifications/Compliance ANSI, SLAS
    Volume (Metric) Well 330μL
    Color Black
    Applications Solid phase with hydrophobic molecules and liquid phase with hydrophilic molecules
    Bottom Diameter F-bottom

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit