Learn More
BrandTech™ BRANDplates lipoGrade™ 96-Well Microplates Available on GSA/VA Contract for Federal Government customers only.
Immunoassay plate optimized for the immobilization of hydrophobic molecules
Manufacturer:  BrandTech™ 781843
 View more versions of this product
Catalog No. 13-882-237
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
C-bottom
Transparent
Polystyrene
96-Well Assay Microplate
Round
High
ANSI, SLAS
Solid phase with hydrophobic molecules and liquid phase with hydrophilic molecules
No
350μL