BrandTech™ BRANDplates lipoGrade™ 96-Well Microplates Available on GSA/VA Contract for Federal Government customers only.

Featuring a strongly hydrophobic/lipophilic surfacehomogeneous assays with hydrophilic molecules that remain in solution

Manufacturer: BrandTech™ 781849

 View more versions of this product

Catalog No. 13-882-241

  Check Availability
Add to Cart

    Specifications

    Lid No,No
    For Use With (Application) Immobilization of hydrophobic molecules
    Format 96-well,96-well
    Material Polystyrene,Polystyrene
    No. of Wells 96
    Sterile No
    Type 96-Well Microplate,96-Well Microplate
    Volume (Metric) 350μL,350μL
    Certifications/Compliance ANSI, SLAS
    Well Design Round
    Closure No
    Color Black
    Well Shape Round
    Wells 96

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit