Learn More
BrandTech™ BRAND™ macro™ Pipet Controller Accessories Available on GSA/VA Contract for Federal Government customers only.
Allows continued use after years of use. BrandTech™ BRAND™ macro™ Pipet Controller Accessories available for order include Suction Bellows, Valve System, Non-sterile 3µ Membrane Filters, and Silicone Adapters.
Manufacturer:  BrandTech™ 26056
 View more versions of this product
Catalog No. 21-381-237
  Check Availability
Add to cart
Specifications
Description
Accessories and replacement parts for the BRAND™ macro™ Pipet Controller include
  • Suction Bellows: Just a simple squeeze of the suction bellows powers the aspiration of up to 50mL. Need to fill a 100mL pipet? Just squeeze the bellows again and continue! The bellows are replaceable, if necessary, after years of use.
  • Unique Valve System: Wide air channels allow fast filling of large pipets. Sensitive valves give precise control of low-volume pipets. The replaceable valve system is designed to ensure appropriate drain times for accurate measurement.
  • Non-sterile 3µ Membrane Filters: Available in packs of 10.
  • Silicone Adapter: Detachable and autoclavable, and fits 0.1 to 200mL pipets. Hydrophobic filter limits over aspiration into unit.
Description & Specifications
Specifications
No
BrandTech™ BRAND™ macro™ Pipet Controller
3μm
3μ Membrane Filter
Polypropylene, PTFE