Learn More
BrandTech™ BRAND™ nano-cap™ Pipet Tips
Manufactured from high purity, DiHEMDA- and oleamide-free, virgin polypropylene. BrandTech™ BRAND™ nano-cap™ Pipet Tips provide clean, hydrophobic surfaces, reduce sample retention, and increase accuracy and reproducibility.
Manufacturer:  BrandTech™ 702312
 View more versions of this product
Catalog No. 13-688-604
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene.
  • Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
  • Available either in bulk packages or in convenient rack-and-stack formats
Description & Specifications
Specifications
Refill Tip
20, 100μL
Polypropylene
960 Pack
0.1 to 20μL
Nano-cap™
TipRefill: 10 filled 96-tip inserts
DiHEMDA- and oleamide-free