BrandTech™ BRAND™ Pipet Tips, 1 to 50uL Available on GSA/VA Contract for Federal Government customers only.

Fits most pipettes that use standard ultra-micro/crystal tips

Manufacturer: BrandTech™ 732306

 View more versions of this product

Catalog No. 13-889-192

  Check Availability
Add to Cart

    Specifications

    Product Type Tip
    Material Polypropylene
    Sterility Nonsterile
    Autoclavable Yes
    Tip Style Ultra Low Retention
    Packaging Rack
    Quantity 480 Pack
    Certifications/Compliance DNA, RNase, ATP and endotoxin-free
    For Use With (Application) High translucency, easy ejection and low waste, quick volume check
    Volume (Metric) 1 to 50μL
    Color Colorless

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit