BrandTech™ BRAND™ Pipet Tips, 2 to 200uL: Standard Retention Available on GSA/VA Contract for Federal Government customers only.

Manufactured from oleamide- and diHEMDA-free high-purity polypropylene. BrandTech™ BRAND™ Pipet Tips, 2 to 200µL: Standard Resolution are thin-walled for high translucency, easy ejection, and low waste. Graduations marked at 20 and 100µL for quick volume check.All unopened packages are DNA-, RNase-, ATP- and endotoxin-free.

Manufacturer: BrandTech™ 732248

 View more versions of this product

Catalog No. 13-889-210

  Check Availability
Add to Cart

    Specifications

    Product Type Tip
    Material Polypropylene
    For Use With (Equipment) For pipets with yellow color code
    Sterility Nonsterile
    Packaging TipStack™, 10 x 96 tips
    Graduations 20, 100μL
    Autoclavable Yes
    Tip Style Standard
    Quantity 960 Pack
    Certifications/Compliance DNA, RNase, ATP and endotoxin-free
    Volume (Metric) 2 to 200μL
    Color Colorless

    • High-purity polypropylene is oleamide- and diHEMDA-free
    • Nonsterile
    • All unopened packages are DNA-, RNase-, ATP- and endotoxin-free
    • Thin-walled for high translucency, easy ejection and low waste
    • Graduations at 20 and 100µL for quick volume check
    • Available either in bulk packages or in convenient, ecological rack-and-stack formats
    • Bulk tips are yellow colored
    • Racked tips are colorless and placed into yellow-colored tip tray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit