Learn More
BrandTech™ BRAND™ Pipet Tips, 2 to 200uL: Ultra-Low Retention Available on GSA/VA Contract for Federal Government customers only.
Low retention tips used for virtually all pipettes with yellow color code
Manufacturer:  BrandTech™ 732348
 View more versions of this product
Catalog No. 13-889-216
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tip
Nonsterile
20, 100μL
Ultra Low Retention
Rack
DNA, RNase, ATP and endotoxin-free
2 to 200μL
Colorless
Yes
Polypropylene
960 Pack
For pipettes with yellow color code