Learn More
BrandTech™ BRAND™ Pipet Tips, 2 to 200uL: Ultra-Low Retention Available on GSA/VA Contract for Federal Government customers only.
Low retention tips used for virtually all pipettes with yellow color code
Manufacturer:  BrandTech™ 732308
 View more versions of this product
Catalog No. 13-889-193
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tip
Nonsterile
2, 10μL
Ultra Low Retention
Rack
DNA, RNase, ATP and endotoxin-free
0.5 to 10μL
Colorless
Yes
Polypropylene
960 Pack
High translucency, easy ejection and low waste, quick volume check