BrandTech™ BRAND™ Pipet Tips, 5 to 300uL Available on GSA/VA Contract for Federal Government customers only.

Used for virtually all pipettes with yellow color code. BrandTech™ Pipet Tips, 5 to 300µL are manufactured from oleamide- and diHEMDA-free high-purity polypropylene and are available in either standard or ultra low retention types. Thin-walled for high translucency, easy ejection, and low waste. Graduations marked at 50, 100 and 300µL for quick volume check.

Manufacturer: BrandTech™ 732110

 View more versions of this product

Catalog No. 13-889-156

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Tip
    Material Polypropylene
    Length (English) 2.08 in.
    Length (Metric) 5.3cm
    Sterility Nonsterile
    Packaging TipRefill: 10 Refills x 96 each
    Graduations 50, 100, 300μL
    Tip Style Standard
    Quantity 960 Pack
    Certifications/Compliance DNA, RNase, ATP and endotoxin-free
    For Use With (Application) Reverse pipetting, plate washing, working with multichannel pipettes
    Volume (Metric) 5 to 300μL
    Color Colorless

    Used for virtually all pipettes with yellow color code.

    • High-purity polypropylene is oleamide- and diHEMDA-free
    • Nonsterile
    • All unopened packages are DNA-, RNAse, ATP- and endotoxin-free
    • Thin-walled for high translucency, easy ejection and low waste
    • Graduation at 50, 100 and 300µL for quick volume check
    • Available in either standard or ultra low retention types
    • Available either in bulk packages or in convenient, ecological rack format
    • Racked tips are colorless and placed into green-colored tip tray

    Ultra low retention tips

    • Produced through special, patented physicochemical process
    • Homogeneous, defect-free surface for low surface tension and minimal sample retention
    • Reduce sample loss and provide substantially higher reproducibility when working with critical media
    • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can increase sample loss
    • High chemical resistance
    • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit