Learn More
BrandTech™ BRAND™ Pipet Tips, 50 to 1000uL Available on GSA/VA Contract for Federal Government customers only.
Manufactured in an environmentally friendly manner using only high-purity polypropylene, free from oleamide and diHEMDA. BrandTech™ BRAND™ Pipet Tips, 50 to 1000µL are thin-walled for high translucency, easy ejection, and low waste. Graduations provide quick volume assessment.
Manufacturer:  BrandTech™ 732252
 View more versions of this product
Catalog No. 13-889-211
  Check Availability
Add to cart
Specifications
Description
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Nonsterile
  • All unopened packages are DNA-, RNase-, ATP- and endotoxin-free
  • Thin-walled for high translucency, easy ejection and low waste
  • Graduations at 250, 500 and 1000µL for quick volume check
  • Available either in bulk packages or in convenient, ecological rack-and-stack formats
  • Bulk tips are blue colored and racked tips are colorless and placed into blue-colored tip tray
Description & Specifications
Specifications
Tip
Nonsterile
250, 500, 1000μL
Polypropylene
960 Pack
DNA, RNase, ATP and endotoxin-free
50 to 1000μL
Colorless
Standard
Racked, TipStack, 2 x 5 racks of 96 pieces
7cm
For routine laboratory and research pipetting