Learn More
BrandTech™ BRAND™ Pipet Tips, 50 to 1000uL: Ultra-Low Retention Available on GSA/VA Contract for Federal Government customers only.
Ultra-low retention tips for routine laboratory and research pipetting
Manufacturer:  BrandTech™ 732312
 View more versions of this product
Catalog No. 13-889-195
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tip
Nonsterile
2, 10μL
Ultra Low Retention
Rack
DNA, RNase, ATP and endotoxin-free
0.5 to 10μL
Colorless
Yes
Polypropylene
960 Pack
For routine laboratory and research pipetting