BrandTech™ BRAND™ Ultra-micro Pipet Tips
Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene
Manufacturer: BrandTech™ 702526
 View more versions of this product
Catalog No. 13-688-605
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Tip
    Material Polypropylene
    Sterility Nonsterile
    Graduations 20, 100μL
    Tip Style Ultra micro
    Packaging Bulk
    Quantity 2000 Pack
    Certifications/Compliance DiHEMDA- and oleamide-free
    For Use With (Application) For clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility
    Volume (Metric) 0.5 to 20μL
  • Description & Specifications
Related Products
Product Content Feedback