BrandTech™ BRAND™ White 96-well PCR Plates Available on GSA/VA Contract for Federal Government customers only.

Designed specifically to optimize results when performing quantitative Real Time PCR (qPCR)

Manufacturer: BrandTech™ 781354

 View more versions of this product

Catalog No. 13-878-205

  Check Availability
Add to Cart

    Specifications

    Lid No,No
    For Use With (Application) Optimize results when performing quantitative RT-PCR
    Format 96-well,96-well
    Includes No Skirt
    Material Polystyrene,Polystyrene
    No. of Wells 96
    Sterile No
    Type 96-Well PCR Plate,96-Well PCR Plate
    Volume (Metric) 0.2mL,0.2mL
    Closure No
    Color White
    Wells 96

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit