BrandTech™ BRANDplates™ cellGrade™ premium 384-Well Microplates Available on GSA/VA Contract for Federal Government customers only.

For serum-free and serum-reduced cultivation of cells

Manufacturer: BrandTech™ 782100

 View more versions of this product

Catalog No. 14-386-055

  Check Availability
Add to Cart

    Specifications

    Lid Yes,Yes
    For Use With (Application) Cultivation of fastidious adherent cell lines
    Format 384-well,384-well
    Material Polystyrene,Polystyrene
    No. of Wells 384
    Sterile Yes
    Surface Coating Carboxyl, Hydroxyl and free Amino groups
    Type Premium 384 -Well Standard Microplate,Premium 384 -Well Standard Microplate
    Volume (Metric) 100μL,100μL
    Certifications/Compliance ANSI, SLAS
    Volume (Metric) Well 100μL,100μL
    Well Design Flat (F)
    Closure Yes
    Color Transparent
    Wells 384

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit