Learn More
BrandTech™ BRANDplates™ hydroGrade™ 384-Well Microplates Available on GSA/VA Contract for Federal Government customers only.
Immunoassay plate optimized for immobilization of hydrophilic molecules
Manufacturer:  BrandTech™ 781800
 View more versions of this product
Catalog No. 14-386-033
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • High affinity for molecules with hydrophilic regions such as glycoproteins, peptides and more
  • PS, nonsterile
  • For solid phase assays with hydrophilic molecules and liquid phase with hydrophobic molecules, respectively
  • Alternative to immunoGrade™ surface
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase, noncytotoxic
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Flat
For homogeneous assays with hydrophobic molecules that remain in solution
0.1mL
Non-sterile
Clear
Polystyrene
384-Well Microplate