Learn More
BrandTech™ BRANDplates™ hydroGrade™ 384-Well Microplates
Immunoassay plate optimized for immobilization of hydrophilic molecules
Manufacturer:  BrandTech™ 781802
 View more versions of this product
Catalog No. 14-386-035
Add to cart
Description
Description
  • High affinity for molecules with hydrophilic regions such as glycoproteins, peptides and more
  • PS, nonsterile
  • For solid phase assays with hydrophilic molecules and liquid phase with hydrophobic molecules, respectively
  • Alternative to immunoGrade™ surface
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase, noncytotoxic
Specifications
Specifications
Flat
For homogeneous assays with hydrophobic molecules that remain in solution
0.1mL
Non-sterile
Black
Polystyrene
384-Well Microplate