Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRANDplates™ Insert Strips With Inlet Opening System

Available with two membranes PC or PET membrane

Manufacturer:  BrandTech™ 782871

 View more versions of this product

Catalog No. 14-380-862


Add to cart

Description

Description

  • Sterile
  • cellGrade™ plus surface
  • With inlet channels that enables enables rapid, consistent changing of media, from submersion culture to air-lift culture
  • Inlet channels enables adjustment of medium level without damage to tissue model
  • Membrane material: PC or PET material for different optical requirements and cell visibility
  • Multiple membrane pore sizes for different applications: 0.4µm, 1.0µm, 3.0µm, 8.0µm
  • 12 strips/Pk. of four wells each

Applications:

Automated in vitro preparation of human tissues

Specifications

Specifications

PET membrane
12/Pk.
Insert Strips With Inlet Opening System
8μm
cellGrade™ plus
Sterile
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit