Learn More
BrandTech™ BRANDplates™ lipoGrade™ 384-Well Microplates
Immunoassay plate optimized for the immobilization of hydrophobic molecules
Manufacturer:  BrandTech™ 781862
 View more versions of this product
Catalog No. 14-386-038
Add to cart
Description
Description
  • High binding for molecules with predominately hydrophobic regions such as lipoproteins or peptides
  • PS, non-sterile
  • For solid phase assays with hydrophobic molecules and liquid phase with hydrophilic molecules, respectively
  • Alternative to immunoGrade™ surface
  • Available in transparent, white or black color
  • With standard or transparent bottom
  • Modification via physical-chemical process, good shelf life at room temperature
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
Specifications
Specifications
Flat
For immobilization of biomolecules with strongly hydrophobic regions
384-Well Microplate
Black
0.1mL
Non-sterile