BrandTech™ BRANDplates™ lipoGrade™ 384-Well Microplates Available on GSA/VA Contract for Federal Government customers only.

Immunoassay plate optimized for the immobilization of hydrophobic molecules

Manufacturer: BrandTech™ 781866

 View more versions of this product

Catalog No. 14-386-042

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) For immobilization of biomolecules with strongly hydrophobic regions
    Sterile No
    Type 384-Well Microplate
    Volume (Metric) Well 0.12mL,0.12mL
    Well Design Flat (F)
    Color White
    Applications For immobilization of biomolecules with strongly hydrophobic regions

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit