Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRANDplates™ Microplate Insert System With Smooth-walled Insert Strips Available on GSA/VA Contract for Federal Government customers only.

Six well plates filled with six insert strips

Manufacturer:  BrandTech™ 782812

 View more versions of this product

Catalog No. 14-380-839


  Check Availability
Add to cart

Specifications

Description

  • Sterile
  • With smooth-walled insert strips
  • cellGrade™ plus surface
  • Membrane material: PC or PET material
  • Multiple membrane pore sizes for different applications: 0.4µm, 1.0µm, 3.0µm, 8.0µm
  • 5 plates/Pk. with lids and 30 insert strips

Applications:

in vitro preparation of tissue (3D tissue culture)

Description & Specifications

Specifications

PET membrane
0.4μm
Yes
Microplate Insert System w/Insert Strips
6
5/Pk.
cellGrade™ plus
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit