BrandTech™ BRANDplates™ Microplate Insert System With Smooth-walled Insert Strips
Six well plates filled with six insert strips
Manufacturer: BrandTech™ 782852
 View all options for this product
Catalog No. 14-380-855
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Membrane PET membrane
    No. of Wells 6
    Pore Size 3μm
    Quantity 5/Pk.
    Sterile Yes
    Surface Coating cellGrade™ plus
    Type Microplate Insert System w/Insert Strips
    • Sterile
    • With smooth-walled insert strips
    • cellGrade™ plus surface
    • Membrane material: PC or PET material
    • Multiple membrane pore sizes for different applications: 0.4µm, 1.0µm, 3.0µm, 8.0µm
    • 5 plates/Pk. with lids and 30 insert strips
    in vitro preparation of tissue (3D tissue culture)
  • Description & Specifications
Related Products
Product Content Feedback