BrandTech™ BRANDplates™ Microplate Insert System With Smooth-walled Insert Strips Available on GSA/VA Contract for Federal Government customers only.

Six well plates filled with six insert strips

Manufacturer: BrandTech™ 782802

 View more versions of this product

Catalog No. 14-380-835

  Check Availability
Add to Cart

    Specifications

    Pore Size 0.4μm
    Quantity 5/Pk.
    Sterile Yes
    Membrane PC membrane
    No. of Wells 6
    Surface Coating cellGrade™ plus
    Type Microplate Insert System w/Insert Strips,Microplate Insert System w/Insert Strips
    Wells 6

    • Sterile
    • With smooth-walled insert strips
    • cellGrade™ plus surface
    • Membrane material: PC or PET material
    • Multiple membrane pore sizes for different applications: 0.4µm, 1.0µm, 3.0µm, 8.0µm
    • 5 plates/Pk. with lids and 30 insert strips

    Applications:

    in vitro preparation of tissue (3D tissue culture)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit