Learn More
BRAND™ BRANDplates™ pureGrade™ 384-Well Microplates
Non-treated microplates for diverse applications
Supplier: BRAND™ 781622
Description
- Non-treated PS surface
- Medium binding, non-sterile
- Standard plate for most applications
- Free from endotoxins, DNase, DNA, RNase; noncytoxic
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
384 | |
Non-treated | |
For homogenous assays, screenings, storage |
Without Lid | |
Black | |
0.1 mL | |
Non-sterile | |
Flat Bottom (F-well) | |
50/Cs. |