BrandTech™ BRANDplates™ pureGrade™ 96-Well Microplates Available on GSA/VA Contract for Federal Government customers only.

Constructed of polystyrene with nontreated surface. BrandTech™ BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. 
Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.

Manufacturer: BrandTech™ 781602

 View more versions of this product

Catalog No. 13-882-202

  Check Availability
Add to Cart

    Specifications

    Barcode Yes
    For Use With (Application) Homogenous assays, screenings and storage
    Binding Property Medium
    Material Polystyrene
    Sterile No
    Type 96-Well Assay Microplate
    Certifications/Compliance ANSI, SLAS
    Volume (Metric) Well 350μL
    Well Design Flat (F),Flat (F)
    Color Transparent
    Applications Homogenous assays, screenings and storage

    • Polystyrene surface
    • Medium binding, nonsterile
    • Standard plate for most applications
    • Gray alphanumeric embossing on white and black solid plates for easy well identification
    • Strip plates available
    • Standard plates available in U-, V-, F-, C-shapes
    • Transparent bottoms available in F- shape
    • Available in clear, white or black
    • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
    • Suited for homogenous assays, screening, and storage.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit