BrandTech™ BRANDplates™ pureGrade™ S 1536-well Microplates Available on GSA/VA Contract for Federal Government customers only.

Non-treated sterile microplates for diverse applications

Manufacturer: BrandTech™ 781700

 View more versions of this product

Catalog No. 14-386-006

  Check Availability
Add to Cart

    Specifications

    Lid Yes
    Type 1536-Well Microplate
    Color Transparent
    For Use With (Application) Sterile applications, bacteriological assays
    Bottom Flat (F)
    Sterile Yes
    Certifications/Compliance ISO 11137, AAMI
    Volume (Metric) Well 10μL,10μL
    Closure Yes,Yes
    Well Bottom Flat (F)
    Applications Sterile applications, bacteriological assays

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit