Learn More
BrandTech™ BRANDplates™ pureGrade™ S 1536-well Microplates
Non-treated sterile microplates for diverse applications
Manufacturer:  BrandTech™ 781702
 View more versions of this product
Catalog No. 14-386-008
Add to cart
Description
Description
  • Nontreated PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Sterile plate for most applications
  • With lid
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Bacteriological assays, screenings, storage.
Specifications
Specifications
Flat
Black
10μL
1536-Well Cell Culture Multiwell Plates
ISO 11137, AAMI
Sterile applications, bacteriological assays
With Lid
Sterile