Learn More
BRAND™ BRANDplates™ pureGrade™ S 384-well Microplates
Non-treated sterile microplates for diverse applications
Supplier: BRAND™ 781681
Description
- Non-treated PS surface
- Medium binding
- Sterilized via beta-radiation
- Sterile plate for most applications
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
384 | |
Non-treated | |
Laboratory tasks, such as homogeneous assays, screening and storage |
With Lid | |
White | |
0.1 mL | |
Sterile | |
Flat Bottom (F-well) | |
50/Cs. |