BrandTech™ BRANDplates™ pureGrade™ S 96-well Microplates Available on GSA/VA Contract for Federal Government customers only.

Nontreated sterile microplates for diverse applications

Manufacturer: BrandTech™ 781670

 View more versions of this product

Catalog No. 13-882-216

  Check Availability
Add to Cart

    Specifications

    Lid Yes
    Barcode Yes
    For Use With (Application) Bacteriological assays, screening and for storage
    Binding Property Medium
    Material Polystyrene
    Sterile Yes
    Type 96-Well Assay Microplate
    Certifications/Compliance ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS
    Volume (Metric) Well 330μL
    Well Design Flat (F),Flat (F)
    Closure Yes
    Color White/Transparent Bottom
    Applications Bacteriological assays, screening and for storage

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit