Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ Class A PMP Volumetric Flasks with NS Stoppers Available on GSA/VA Contract for Federal Government customers only.

These Class A Volumetric Flasks with NS Stoppers, calibrated “to contain”, are an excellent break-resistant alternative to glass in the laboratory.

Manufacturer:  BrandTech™ V67304

 View more versions of this product

Catalog No. 14-379-222


Add to cart

Description

Description

  • Meet Class A tolerances per DIN EN ISO 1042
  • Manufactured from crystal-clear polymethylpentene
  • Supplied certificate includes mean test value and lot (lot and year imprinted on product)
  • Thermal stress up to 121°C (autoclaving) does not cause permanent exceeding of the tolerance limits
  • However, to preserve markings, cleaning at no higher than 60°C is recommended
  • Available in a range of capacities and stopper sizes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Yes
Polypropylene
Translucent
PMP
Imprinted lot number, batch certificate
14/23
0.1mL
Calibrated "to contain" per DIN EN ISO 1042
NS Stopper
100 mL
180mm
6/pk
121°C max.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit