Learn More
BrandTech™ Digital Dispensette™ Organic Bottle-Top Dispensers Available on GSA/VA Contract for Federal Government customers only.
Ideal for HPLC and combinatorial chemistry labs
Manufacturer:  BrandTech™ 4731350
 View more versions of this product
Catalog No. 13-642-888
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Bottletop Dispenser
Up to 2.2g/cm3
Yes
Up to 600mbar
Discharge tube, 125 to 240mm/170 to 330mm L telescoping filling tube, valve mounting/calibration tool, 28, 33 and 38mm thread polypropylene adapters, certificate of performance and operating manual
15° to 40°C
0.084 to 0.845 oz.
Up to 500mm2/sec.
Digital
≤0.5% ±125μL
Dispensing organic solvents including chlorinated andfluorinated hydrocarbons, concentrated acids , trifluoroacetic acid , tetrahydrofuran , methanol, acetonitrile, methylene chloride, DMF and DMSO
0.1mL
≤0.1% 25μL
59° to 104°F
2.5 to 25mL
Accessories