Learn More
BrandTech™ Flexible Discharge Tubes for Dispensette™ III Bottle-Top Dispensers Available on GSA/VA Contract for Federal Government customers only.
Enhances the suitability of Dispensette dispensers for various applications.
Manufacturer:  BrandTech™ 707926
 View more versions of this product
Catalog No. 13-688-75
  Check Availability
Add to cart
Specifications
Description
  • For serial dispensing such as scintillation cocktail, the flexible discharge tube preserves all important safety innovations
  • PTFE material
  • Fine tip
Order Info
Not for use with the Dispensette HF or dispensers from other manufacturers.
Description & Specifications
Specifications
Flexible Discharge Tube
Serial dispensing such as scintillation cocktail, the flexible discharge tube preserves all important safety innovations
31.5 in.
800mm
25 to 100mL Dispensette™ III, Dispensette™ organic bottletop dispenser
Safety handle
PTFE
Fine Tip