Learn More
BrandTech™ inertGrade™ 96-well Microplates
Especially suited for cell cultures when adhesion is not desired. Reduction of cell adhesion and protein adsorption minimizes enzyme and cellular activation to aid in the inhibition of the differentiation of progenitor or stem cells and promotes the formation of spheroids.
Manufacturer:  BrandTech™ 781904
 View more versions of this product
Catalog No. 14-386-222
Add to cart
Specifications
Specifications
U-Bottom
Polystyrene
Round
White
330μL