Learn More
BrandTech™ Light-Shielding Class A Volumetric Flasks with PMP Screw Caps
Manufacturer:  BrandTech™ V673040
 View more versions of this product
Catalog No. 14-379-371
Add to cart
Specifications
Specifications
Yes
3.38 oz.
GL18
100 mL
7 in.
Imprinted lot number, batch certificate
Calibrated To Contain
Polypropylene
Individually calibrated ring-marking
PMP
18cm
0.1mL