BrandTech™ NowPak Remote Dispensing System for Dispensette™ III Bottle-Top Dispensers
Manufacturer: BrandTech™ 704284
 View more versions of this product
Catalog No. 13-878--87
  Check Availability
Add to Cart
Alert
Not for use with hydrofluoric acid (HF), peroxides or carbon disulfide. 20-liter NOWPak containers require a longer discharge tube (sold separately).
    Specifications
    Product Type Remote Dispensing System
    For Use With (Equipment) Dispensette™ III and Dispensette™ organic bottletop dispenser
    Includes Filling tube adapter, wall-mounting unit, FEP tubing, adapter for direct mounting, closure cap, clamp seal and drum adapter
  • Description & Specifications
Related Products
Product Content Feedback