Learn More
BrandTech™ PCR 8-Strip Caps Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples. BrandTech™ PCR 8-Strip Caps are autoclavable and RNase-, DNase-, and DNA-free. Domed caps are available in a variety of colors for sample identification.
Manufacturer:  BrandTech™ 781344
 View more versions of this product
Catalog No. 13-882-73
  Check Availability
Add to cart
Specifications
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.
  • Available flat or domed
  • The domed caps are available in a variety of colors for sample identification
  • Autoclavable
  • RNase-, DNase-, and DNA-free.
Description & Specifications
Specifications
Yes
Yes
Polypropylene
Cap strip
Blue
Domed
Yes