BrandTech™ PLASTIBRAND™ PD-Tip™ Disposable Pipet Tips Available on GSA/VA Contract for Federal Government customers only.
Made of HDPE and polypropylene (polypropylene and liquid crystal polymer for 0.1mL size) without lubricants or additives that can contaminate samples. BrandTech™ PLASTIBRAND™ PD-Tip™ Disposable Pipet Tips are automatically recognized by HandyStep™ electronic and AutoRep E. Bio-Cert™ tips are independently certified free from DNA, RNase, ATP, and endotoxin for critical biological applications.
Manufacturer: BrandTech™ 702374
 View more versions of this product
Catalog No. 13-688-364
  Check Availability
Add to Cart
    Specifications
    Material HDPE/Polypropylene
    For Use With (Equipment) BRAND Handystep and most standard repeating pipetters
    Sterility Nonsterile
    Volume (Metric) 2.5mL
    • Made of high-density polyethylene and polypropylene (polypropylene and liquid crystal polymer for 0.1mL size) without lubricants or additives that can contaminate samples.
    • Tip size is automatically recognized by HandyStep™ electronic and AutoRep E.
    • Available in nonsterile and Bio-Cert™ sterile. Bio-Cert™ tips are independently certified free from DNA, RNase, ATP, and endotoxin for critical biological applications.
  • Description & Specifications
Related Products
Product Content Feedback