Learn More
BrandTech™ Polypropylene Test Tube Racks Available on GSA/VA Contract for Federal Government customers only.
Constructed of sturdy polypropylene. BrandTech™ Polypropylene Test Tube Racks include alphanumeric positions for easy sample identification. Racks are stackable, non-conductive and will not float in water baths. Suitable for use in temperatures from -20° C to 90° C and supplied in three pieces for easy and permanent assembly.
Manufacturer:  BrandTech™ 4340033
 View more versions of this product
Catalog No. 03-840-245
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
No
10.43 x 4.96 x 3.46 in. (26.5 x 12.6 x 8.8cm)
8.8cm
10.43 in.
Polypropylene
Rack, Tube
12.6cm
Yellow
3.46 in.
25mm Tubes
26.5cm
32.0
4.96 in.