Learn More
BrandTech™ Botletop Dispenser Replacement Discharge Tube Available on GSA/VA Contract for Federal Government customers only.
Replaces discharge tube in 5, 10mL Dispensette™ III Bottletop Dispenser, 5, 10mL Dispensette™ Organic Bottletop Dispenser, and 25, 50, 100mL Dispensette™ Organic Bottletop Dispenser. BrandTech™ Bottletop Dispenser Replacement Discharge valve includes a viewing window, hinged screw cap swings out of the way when dispensing, and is available with or without a recirculation valve.
Manufacturer:  BrandTech™ 707936
 View more versions of this product
Catalog No. 02-681-441
  Check Availability
Add to cart
Description
Description
  • Viewing window provides easy priming verification
  • Hinged screw cap swings out of the way when dispensing
  • Discharge valve with safety ball closes when discharge valve is not mounted
  • Available with or without recirculation valve
For use with
  • 5, 10mL Dispensette&trade III Bottletop Dispenser
  • 5, 10mL Dispensette&trade Organic Bottletop Dispenser
  • 25, 50, 100mL Dispensette&trade Organic Bottletop Dispenser
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yellow
5, 10mL Dispensette™ organic bottletop dispenser
3.54 in.
Standard
Discharge Tube
Integrated valve
90mm