Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BrandTech™ Replacement Parts for Dispensette™ Organic Bottle Top Dispensers: SafetyPrime™ Recirculation Valve
For use with Dispensette Organic Bottle Top Dispenser.
Supplier: BrandTech™ 706090
Includes: Seal and recirculation tube
Description
- For use with Dispensette Organic Bottle Top Dispenser
- Color: Yellow
Specifications
SafetyPrime Recirculation Valve | |
Conserve reagent | |
1/Pk. |
Yellow | |
Dispensette™ organic bottletop dispenser |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction