Learn More
BrandTech™ BRAND™ Reagent Reservoirs Available on GSA/VA Contract for Federal Government customers only.
Constructed of heavy-gauge polypropylene for long-lasting durability. BrandTech™ BRAND™ Reagent Reservoirs are autoclavable and suitable for all 8- and 12-channel pipetters. Non-sterile version includes lid.
Manufacturer:  BrandTech™ 703409
 View more versions of this product
Catalog No. 13-689-207
  Check Availability
Add to cart
Specifications
Description
  • Heavy-gauge polypropylene
  • Suitable for all 8- and 12-channel pipetters
  • Autoclavable
  • Non-sterile version includes lid
Description & Specifications
Specifications
Yes
2.02 oz.
Polypropylene
Yes
For use with multichannel pipets. Without lid.
60mL
40 Bags of 5
Reagent reservoir