Learn More
BrandTech™ BRAND™ Self-adhesive Plate Sealing Film Available on GSA/VA Contract for Federal Government customers only.
Securely seals multiwell plates. BrandTech™ BRAND™ Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate. Sealing films are available in a variety of materials especially suited for storage, immunoassay, or cell and tissue culture.
Manufacturer:  BrandTech™ 781380
 View more versions of this product
Catalog No. 13-883-08
  Check Availability
Add to cart
Specifications
Description
  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
Description & Specifications
Specifications
Aluminum
Cold storage
Roll of 100 Sheets
Sealing film