Learn More
BrandTech™ Technical Grade PFA Wash Bottles
Manufacturer:  BrandTech™ V108792
 View more versions of this product
Catalog No. 14-379-084
Add to cart
Specifications
Specifications
8 oz., 250mL
8 oz.
Yes
250 mL
PFA
Narrow