Learn More
BrandTech™ Transferpette™ S Multichannel Pipet Available on GSA/VA Contract for Federal Government customers only.
Protects against accidental volume changes using innovative one-handed volume adjustment with brake. BrandTech™ Transferpette™ S Multichannel Pipet is supplied with performance certificate, operating manual, shelf mount, two tip boxes, reagent reservoir, pack of replacement v-rings, and one year warranty. Patented individually removable/replaceable tip cones provide easy in-lab cleaning.
Manufacturer:  BrandTech™ 703720
 View more versions of this product
Catalog No. 03-837-306
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±1.6%
9.5 x 5.62 x 4.87 in. (241.3 x 142.8 x 123.8mm)
4.87 in.
Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual
241.3mm
1 Year
142.8mm
0.5 to 10μL
Yes
Digital
123.8mm
9.5 in.
12.5mm
5.62 in.
12