BrandTech™ Transferpette™ S Multichannel Pipet Available on GSA/VA Contract for Federal Government customers only.

Protects against accidental volume changes using innovative one-handed volume adjustment with brake. BrandTech™ Transferpette™ S Multichannel Pipet is supplied with performance certificate, operating manual, shelf mount, two tip boxes, reagent reservoir, pack of replacement v-rings, and one year warranty. Patented individually removable/replaceable tip cones provide easy in-lab cleaning.

Manufacturer: BrandTech™ 703700

 View more versions of this product

Catalog No. 03-837-326

  Check Availability
Add to Cart

    Specifications

    Accuracy ±1.6%
    Autoclavable Yes
    Dimensions (L x W x H) 9.5 x 5.62 x 4.87 in. (241.3 x 142.8 x 123.8mm)
    Display Digital
    Height (English) 4.87 in.
    Height (Metric) 123.8mm
    Includes Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual
    Length (English) 9.5 in.
    Length (Metric) 241.3mm
    Stroke (Metric) Length 12.5mm
    Warranty 1 Year
    Width (English) 5.62 in.
    Width (Metric) 142.8mm
    Number of Channels 8
    Volume (Metric) 0.5 to 10μL

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit