BrandTech™ Transferpette™ S Adjustable Single Channel Pipet Starter Sets Available on GSA/VA Contract for Federal Government customers only.
Simple one-handed operation units exibits high degree accuracy, precision, maximum versatility and reliable performance with Easy Calibration™ technology
Manufacturer: BrandTech™ 704793
 View more versions of this product
Catalog No. 03-837-793
  Check Availability
Add to Cart
    Specifications
    Autoclavable Yes
    For Use With (Application) Demanding laboratory applications
    Includes D-10, D-100, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
    Product Type Standard Set
    Volume (Metric) 0.5 to 10, 10 to 100, 100 to 1000μL
    Type Standard Set
    • Purchase a BrandTech™ Transferpette™ S Adjustable Single Channel Pipette Starter Set and get a complimentary BrandTech Transferpette™ S Multichannel (8-channel) Pipette. To take advantage of this offer, apply for the NLSU program.
      Buy Now
      Expires 2018-06-30
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback