BrandTech™ Transferpette™ S Single Channel Adjustable Pipet Available on GSA/VA Contract for Federal Government customers only.
Exibits a high degree of accuracy, precision, maximum versatility, and reliable performance with Easy Calibration™ technology. BrandTrade; Transferpette™ S Single Channel Adjustable Pipet offers simple one-handed operation with an adjustable brake to protect against accidental volume changes.
Manufacturer: BrandTech™ 704770
 View more versions of this product
Catalog No. 03-837-318
  Check Availability
Add to Cart
    Specifications
    Includes Performance certificate, operating manual, shelf mount, and three year warranty.
    Product Type Single channel pipet
    Accuracy ±1%
    Autoclavable Yes
    Certifications/Compliance CE, IVD
    Compatible Tips 0.1 to 20μL Nano-cap™ tip, 0.5 to 20μL ultra micro tip, 0.1 to 1μL Nano-cap™ tip, 0.5 to 10μL tip
    Display Digital
    Finger Style Curved
    For Use With (Application) Demanding laboratory applications
    For Use With (Equipment) 0.1 to 20μL Nano-cap™ tip, 0.5 to 20μL ultra micro tip, 0.1 to 1μL Nano-cap™ tip, 0.5 to 10μL tip
    Increments 0.01μL
    Warranty 3 Years
    Model D-10
    Volume (Metric) 0.5 to 10μL
    Compatibility 0.1 to 20μL Nano-cap™ tip, 0.5 to 20μL ultra micro tip, 0.1 to 1μL Nano-cap™ tip, 0.5 to 10μL tip
    The Transferpette™ S Single Channel Adjustable Pipet offers robust yet lightweight construction, simple, one-handed operation, a high degree of accuracy and precision with Easy Calibration™ technology for long lasting reliability. The single channel model is available in fixed volume and digital models in volume ranges from 0.1µL to 10mL.
    • Centralized pipetting button with optimized stroke length
    • Adjustable brake to fix volume range
    • Visible four-digit volume display
    • Compatible with universal tips (0.1-1µL requires nano-cap tips)
    • Repeatedly autoclavable at 121°C/250°F(20min.)
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback