BrandTech™ Transferpette™ S Single Channel Adjustable Pipet Available on GSA/VA Contract for Federal Government customers only.
Exibits a high degree of accuracy, precision, maximum versatility, and reliable performance with Easy Calibration™ technology. BrandTrade; Transferpette™ S Single Channel Adjustable Pipet offers simple one-handed operation with an adjustable brake to protect against accidental volume changes.
Manufacturer: BrandTech™ 704772
 View more versions of this product
Catalog No. 03-837-319
  Check Availability
Add to Cart
    Specifications
    Includes Performance certificate, operating manual, shelf mount, and three year warranty.
    Product Type Single channel pipet
    Accuracy ±0.8%
    Autoclavable Yes
    Certifications/Compliance CE, IVD
    Display Digital
    Finger Style Curved
    For Use With (Equipment) 2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip
    Increments 0.02μL
    Warranty 3 Years
    Volume (Metric) 2 to 20μL
    Compatible Tips 2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip
    For Use With (Application) Demanding laboratory applications
    Model D-20
    Compatibility 2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip
    The Transferpette™ S Single Channel Adjustable Pipet offers robust yet lightweight construction, simple, one-handed operation, a high degree of accuracy and precision with Easy Calibration™ technology for long lasting reliability. The single channel model is available in fixed volume and digital models in volume ranges from 0.1µL to 10mL.
    • Centralized pipetting button with optimized stroke length
    • Adjustable brake to fix volume range
    • Visible four-digit volume display
    • Compatible with universal tips (0.1-1µL requires nano-cap tips)
    • Repeatedly autoclavable at 121°C/250°F(20min.)
  • Description & Specifications
Related Products
Product Content Feedback