BrandTech™ Transferpette™ S Single Channel Adjustable Pipet Available on GSA/VA Contract for Federal Government customers only.
Exibits a high degree of accuracy, precision, maximum versatility, and reliable performance with Easy Calibration™ technology. BrandTrade; Transferpette™ S Single Channel Adjustable Pipet offers simple one-handed operation with an adjustable brake to protect against accidental volume changes.
Manufacturer: BrandTech™ 704780
 View more versions of this product
Catalog No. 03-837-322
  Check Availability
Add to Cart
    Specifications
    Includes Performance certificate, operating manual, shelf mount, and three year warranty.
    Product Type Single channel pipet
    Accuracy ±0.6%
    Autoclavable Yes
    Certifications/Compliance CE, IVD
    Compatible Tips 50 to 1000μL tip, 50 to 1250μL tip, 50 to 1000μL tip
    Display Digital
    Finger Style Curved
    For Use With (Application) Demanding laboratory applications
    For Use With (Equipment) 50 to 1000μL tip, 50 to 1250μL tip, 50 to 1000μL tip
    Increments 1μL
    Warranty 3 Years
    Model D-1000
    Volume (Metric) 100 to 1000μL
    Compatibility 50 to 1000μL tip, 50 to 1250μL tip, 50 to 1000μL tip
    The Transferpette™ S Single Channel Adjustable Pipet offers robust yet lightweight construction, simple, one-handed operation, a high degree of accuracy and precision with Easy Calibration™ technology for long lasting reliability. The single channel model is available in fixed volume and digital models in volume ranges from 0.1µL to 10mL.
    • Centralized pipetting button with optimized stroke length
    • Adjustable brake to fix volume range
    • Visible four-digit volume display
    • Compatible with universal tips (0.1-1µL requires nano-cap tips)
    • Repeatedly autoclavable at 121°C/250°F(20min.)
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback